Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHACTR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556029 Supplier Invitrogen™ Supplier No. PA556029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRTRSLPITI EMLKVPDDEE EEEQTCPSTF SEEMTPTSVI PKLPQCLREE EEKESDSDSE GPIQYRDEED EDESYQSALA NKVKRKDTLA MKLNHRPSE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.

This gene encodes a member of the phosphatase and actin regulator (PHACTR) family. Other PHACTR family members have been shown to inhibit protein phosphatase 1 (PP1) activity, and the homolog of this gene in the mouse has been shown to interact with actin and PP1. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHACTR4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHACTR4
Gene Accession No. Q8IZ21
Gene Alias 3110001B12Rik; AI527228; AW495572; C330013F19Rik; humdy; Kiaa4120; mKIAA4120; N28169; PHACTR4; Phosphatase and actin regulator 4; PPP1R124; PRO2963; Protein Humpty dumpty; protein phosphatase 1, regulatory subunit 124
Gene Symbols PHACTR4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHACTR4. Recombinant protein control fragment (Product #RP-94578).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.