Learn More
Thermo Scientific™ Phadia™ Tissue Transglutaminase Antibodies for Celiac, Inflammatory Bowel and Other Gastrointestinal Diseases
Description
If present in the patient's specimen, antibodies to tTG bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgA and IgG antibodies, respectively, are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
- The EliA Celikey Wells are coated with human recombinant tissue transglutaminase (tTG)
- The higher the response value, the more specific antibodies are present in the specimen.
Specifications
Specifications
Type | EliA™ Celikey IgA well |
For Use With (Application) | Gastrointestinal diseases assay |
Quantity | 4 x 12 Tests |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.