Learn More
Thermo Scientific™ Phadiatop™ ImmunoCAP
Catalog No. 14433835p
Click to view available options
Quantity:
48 Pack
This item is not returnable. View return policy
This item is not returnable. View return policy
An initial, qualitative screening test useful for identifying patients with a high likelihood of an allergic disease (so called atopic individuals).
The Phadiatop test utilizes a mixture of allergens often implicated in causing allergic diseases in adults and children. The test differentiates atopic and non-atopic diseases, the first step taken when testing for atopic allergy. A balanced mixture of relevant allergens react with specific IgE antibodies in the patient's serum, demonstrating the presence of atopic allergy to common allergens.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Phadiatop™ ImmunoCAP™
Quantity 48 Pack
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.