Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phakinin Antibody (OTI4E7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273365R
Description
phakinin Monoclonal specifically detects phakinin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
phakinin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BFSP2 | |
Recombinant protein fragment corresponding to amino acids 184-415 of human phakinin (NP_003562) produced in E.coli. | |
0.1 mL | |
8419.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4E7 | |
Western Blot, Immunohistochemistry | |
beaded filament structural protein 2, phakinin, CP47, CP49, Cytoskeletal protein, 49-kD, MGC142078, phakinin | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction