Learn More
Description
Phalloidin-FITC is a green fluorescent cytoskeleton stain. Binds and labels F-actin. For most fluorescent imaging applications, cells should be fixed and permeablilized. Excitation/emission maxima λ ∽496/516 nm.
We suggest making up a stock solution of this product by dissolving the vial contents in 1.5 mL methanol or DMSO. Please refer to the protocol for further information on using this product.
Specifications
Specifications
| Chemical Name or Material | 7-[(4R)-N5-[[(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-5-yl)amino]thioxomethyl]-4-hydroxy-4-methyl-L-ornithine]phalloidin |
| CAS | 915026-99-2 |
| Quantity | 12 μg |
| Target | Fluorescent Actin Probes |
| Molecular Formula | C56H60N10O15S2 |
| Formula Weight | 1177.26 |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

