Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520024
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520024 Supplier Thermo Scientific Supplier No. PA520024
Add to Cart
Add to Cart

PHAP Polyclonal Antibody

PHAP Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen PHAP
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANP32A
Gene Accession No. O35381, P39687, P49911, P97822, Q5XIE0, Q92688, Q9BTT0, Q9EST5, Q9EST6
Gene Alias 2410015B15Rik; 2810018A15Rik; Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; acidic (leucine-rich) nuclear phosphoprotein 32 family, member D; acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; acidic leucine-rich nuclear phosphoprotein 32 family member A; Acidic leucine-rich nuclear phosphoprotein 32 family member B; acidic leucine-rich nuclear phosphoprotein 32 family member E; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family member E; acidic nuclear phosphoprotein 32 family, member A; acidic nuclear phosphoprotein 32 family, member B; acidic nuclear phosphoprotein pp32; Acidic protein rich in leucines; AI047746; AI326868; Anp32; Anp32a; Anp32b; ANP32D; ANP32E; APRIL; C15orf1; cerebellar leucine rich acidic nuclear protein; cerebellar postnatal development protein 1; Cpd1; hepatopoietin Cn; HPPCn; hypothetical protein LOC509685; I1PP2A; inhibitor-1 of protein phosphatase-2A; LANP; LANPL; LANP-L; LANP-like protein; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; leucine-rich acidic nuclear protein like; MAPM; Mapmodulin; mLANP-L; Pal31; PHAP1; PHAPI; PHAPI2; PHAPI2a; Potent heat-stable protein phosphatase 2A inhibitor I1PP2A; PP32; proliferation related acidic leucine rich protein PAL31; proliferation-related acidic leucine-rich protein PAL31; Putative HLA-DR-associated protein I; putative HLA-DR-associated protein I-2; putative human HLA class II associated protein I; silver-stainable protein SSP29; SSP29; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A; W91701
Gene Symbols ANP32A, ANP32B, ANP32E
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at amino terminus of human PHAP.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10541, 11737, 170724, 25379, 361999, 66471, 67628, 8125, 81611
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.