Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA128342
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA128342 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA128342 Supplier Invitrogen™ Supplier No. PA128342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji cell lysate.

Apoptosis is related to many diseases and development. Caspase-9 plays a central role in cell death induced by a variety of apoptosis activators. Cytochrome c, after released from mitochondria, binds to Apaf-1, which forms an apoptosome that in turn binds to and activate procaspase-9. Activated caspase-9 cleaves and activates the effector caspases (caspase-3, -6 and -7), which are responsible for the proteolytic cleavage of many key proteins in apoptosis. The tumor suppressor putative HLA-DR-associated proteins (PHAPs) were recently identified as important regulators of mitochondrion apoptosis. PHAP appears to facilitate apoptosome-medicated caspase-9 activation and to stimulate the mitochondrial apoptotic pathway. PHAP was also shown to oppose both Ras- and Myc-medicated cell transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHAP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene ANP32A
Gene Accession No. O35381, P39687, P49911, P97822, Q5XIE0, Q92688, Q9BTT0, Q9EST5, Q9EST6
Gene Alias 2410015B15Rik; 2810018A15Rik; Acid nuclear phosphoprotein 32 (leucine rich); acidic (leucine-rich) nuclear phosphoprotein 32 family, member A; acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; acidic (leucine-rich) nuclear phosphoprotein 32 family, member D; acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; acidic leucine-rich nuclear phosphoprotein 32 family member A; Acidic leucine-rich nuclear phosphoprotein 32 family member B; acidic leucine-rich nuclear phosphoprotein 32 family member E; acidic nuclear phosphoprotein 32; acidic nuclear phosphoprotein 32 family member A; acidic nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family member E; acidic nuclear phosphoprotein 32 family, member A; acidic nuclear phosphoprotein 32 family, member B; acidic nuclear phosphoprotein pp32; Acidic protein rich in leucines; AI047746; AI326868; Anp32; Anp32a; Anp32b; ANP32D; ANP32E; APRIL; C15orf1; cerebellar leucine rich acidic nuclear protein; cerebellar postnatal development protein 1; Cpd1; hepatopoietin Cn; HPPCn; hypothetical protein LOC509685; I1PP2A; inhibitor-1 of protein phosphatase-2A; LANP; LANPL; LANP-L; LANP-like protein; Leucine-rich acidic nuclear protein; leucine-rich acidic nuclear protein LANP; leucine-rich acidic nuclear protein like; MAPM; Mapmodulin; mLANP-L; Pal31; PHAP1; PHAPI; PHAPI2; PHAPI2a; Potent heat-stable protein phosphatase 2A inhibitor I1PP2A; PP32; proliferation related acidic leucine rich protein PAL31; proliferation-related acidic leucine-rich protein PAL31; Putative HLA-DR-associated protein I; putative HLA-DR-associated protein I-2; putative human HLA class II associated protein I; silver-stainable protein SSP29; SSP29; vacidic (leucine-rich) nuclear phosphoprotein 32 family, member A; W91701
Gene Symbols ANP32A, ANP32B, ANP32E
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids near the amino terminus of human PHAP (accession no. P39687).
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10541, 11737, 170724, 25379, 361999, 66471, 67628, 8125, 81611
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.