Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PHARM-TROL™ Count Precision Size Standards

Catalog No. 09980236 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
15 μm
15.0 ±0.2 μm
Quantity:
20 x 25 mL Bottles
6 x 25 mL Bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
09-980-236 15 μm 20 x 25 mL Bottles
09-980-235 15.0 ±0.2 μm 6 x 25 mL Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-980-236 Supplier Thermo Scientific™ Supplier No. CSBX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Document the reproducibility of a particle counter by permitting a continuous record of its performance.

Document the reproducibility of a particle counter by permitting a continuous record of its performance with Thermo Scientific™ PHARM-TROL™ Count Precision Size Standards, ready-to-use precision standards designed for regular use in liquid particle counters for USP <788> and <789> verification. Data is documented for internal or customer audits by using a NIST™-traceable microsphere suspension with a known concentration.

Features:

  • Contains NIST™-traceable size standards with a measured particle count
  • Developed for manufacturers of parenteral drugs and ophthalmic solutions seeking interim verification of USP <788> and <789> verification (Particulate Matter in Injections and Particulate Matter in Opthalmic Solutions)
  • Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology
  • Prepared with the same exacting procedures as the USP Count Reference Standard, providing process control data for particle counting consistency and verification of size calibration
TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant
Concentration 3800 particles/mL ± 15%
Content And Storage Unless otherwise stated, refrigerate 2° to 8deg;C product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Contents Polystyrene microspheres in water
Description Used by manufatures of parenteral drugs and ophthalmic solutions who are seeking interim verification of USP <788> and <789>.
Color White or Colorless
For Use With (Application) USP Verification
Material Polystyrene
Product Type Count Control Particles
Includes Certificate of Calibration and Traceability to NIST
Particle Density 1.05g/cm3
Quantity 20 x 25 mL Bottles
Refractive Index 1.59 at 589nm (25°C)
Size Distribution <3%
Stability Product stable for a minimum of 12 months
Diameter (Metric) 15 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.