Learn More
Foamtec PharmaMOP™ Cleanroom Mop Heads, Sterile
Catalog No. p-4370286
Click to view available options
Material:
Microfiber
Polyester
Product Type:
EZ Glide Gamma Irradiated Mop Head
EZ Glide Mop Head
Gamma Irradiated Mop Head
High Fluid Capacity Cleanroom Mop Head
For cleaning and disinfection of rough surfaces and high soil areas
Enables the quick, efficient cleaning of surfaces stained with disinfectant residues
  • Gamma irradiated
  • EzGLIDE™ polyester mop head
  • 0.125 in. foam core
Certifications
ISO 9001:2000
Specifications
Product Type Gamma Irradiated Mop Head
Sterility Sterile
Autoclavable Autoclavable
For Use With (Application) For cleaning and disinfection of rough surfaces and high soil areas, ISO class 1 to 7 cleanrooms
Certifications/Compliance ISO 9001:2000
Material Polyester
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.