Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHAX Antibody, Novus Biologicals™
SDP

Catalog No. NB395977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395977 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395977 Supplier Novus Biologicals Supplier No. NBP24882125UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHAX Polyclonal antibody specifically detects PHAX in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PHAX
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias FLJ13193, phosphorylated adapter RNA export protein, phosphorylated adaptor for RNA export, RNA U small nuclear RNA export adapter protein, RNA U, small nuclear RNA export adapter (phosphorylation regulated), RNA U, small nuclear RNA export adaptor (phosphorylation regulated), RNUXA
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: ESQEHTKDLDKELDEYMHGGKKMGSKEEENGQGHLKRKRPVKDRLGNRPEMNYKGRYEITAEDSQEKVADEISFRLQEPKKDLI
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51808
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.