Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHAX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584426 Supplier Invitrogen™ Supplier No. PA584426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDSDMTVAPS DRPLQLPKVL GGDSAMRAFQ NTATACAPVS HYRAVESVDS SEESFSDSDD DSCLWKRKRQ KC Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 69%.

A phosphoprotein adapter involved in the XPO1-mediated U snRNA export from the nucleus. Bridge components required for U snRNA export, the cap binding complex (CBC)-bound snRNA on the one hand and the GTPase Ran in its active GTP-bound form together with the export receptor XPO1 on the other. Its phosphorylation in the nucleus is required for U snRNA export complex assembly and export, while its dephosphorylation in the cytoplasm causes export complex disassembly. It is recycled back to the nucleus via the importin alpha/beta heterodimeric import receptor. The directionality of nuclear export is thought to be conferred by an asymmetric distribution of the GTP- and GDP-bound forms of Ran between the cytoplasm and nucleus. Its compartmentalized phosphorylation cycle may also contribute to the directionality of export. Binds strongly to m7G-capped U1 and U5 small nuclear RNAs (snRNAs) in a sequence-unspecific manner and phosphorylation-independent manner. Plays also a role in the biogenesis of U3 small nucleolar RNA (snoRNA). Involved in the U3 snoRNA transport from nucleoplasm to Cajal bodies. Binds strongly to m7G-capped U3, U8 and U13 precursor snoRNAs and weakly to trimethylated (TMG)-capped U3, U8 and U13 snoRNAs. Binds also to telomerase RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHAX
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHAX
Gene Accession No. Q9H814
Gene Alias 26 kDa resiniferatoxin-binding protein; 2810055C14Rik; 4933427L19Rik; AU018701; AU018854; cytosolic resiniferatoxin-binding protein; D18Ertd65e; p55; PHAX; Phosphorylated adapter RNA export protein; phosphorylated adaptor for RNA export; phosphorylation regulated; RBP-2; RBP-26; resiniferatoxin-binding protein 2; RNA U small nuclear RNA export adapter protein; RNA U, small nuclear RNA export adapter (phosphorylation regulated); RNA U, small nuclear RNA export adaptor; RNA U, small nuclear RNA export adaptor (phosphorylation regulated); Rnuxa; RTX-42
Gene Symbols PHAX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHAX. Recombinant protein control fragment (Product #RP-105135).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51808
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.