Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHC2 Monoclonal Antibody (1F4)
GREENER_CHOICE

Catalog No. PIMA524404
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524404 Supplier Invitrogen™ Supplier No. MA524404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PYLQESKEEG APLKLKCELC GRVDFAYKFK RSKRFCSMAC AKRYNVGCTK RVGLFHSDRS KLQKAGAATH NRRRASKASL PPLTKDTKKQ PTGTVPLSVT AALQLTHSQE.

In Drosophila melanogaster, the 'Polycomb' group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein 'polyhomeotic' (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHC2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Phc2
Gene Accession No. Q8IXK0
Gene Alias A3galt2; AA415044; D130050K19Rik; D4Ertd810e; early development regulator 2; early development regulator 2 (homolog of polyhomeotic 2); early development regulatory protein 2; Edr2; hPH2; mPH2; p36; Ph2; Phc2; polyhomeotic 2; polyhomeotic 2 protein; polyhomeotic homolog 2; polyhomeotic homolog 2 (Drosophila); polyhomeotic-like 2; polyhomeotic-like 2 (Drosophila); polyhomeotic-like protein 2
Gene Symbols Phc2
Host Species Mouse
Immunogen PHC2 (NP_004418.2, 91 a.a. ~ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1912
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.