Learn More
Invitrogen™ PHC2 Polyclonal Antibody

Description
Immunogen sequence: HFLPSEPTKW NVEDVYEFIR SLPGCQEIAE EFRAQEIDGQ ALLLLKEDHL MSAMNIKLGP ALKIYARISM LKDS; Positive Samples: DU145, HT-29, HeLa, Jurkat, 293T; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | PHC2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Phc2 |
| Gene Accession No. | Q8IXK0 |
| Gene Alias | A3galt2; AA415044; D130050K19Rik; D4Ertd810e; early development regulator 2; early development regulator 2 (homolog of polyhomeotic 2); early development regulatory protein 2; Edr2; hPH2; mPH2; p36; Ph2; Phc2; polyhomeotic 2; polyhomeotic 2 protein; polyhomeotic homolog 2; polyhomeotic homolog 2 (Drosophila); polyhomeotic-like 2; polyhomeotic-like 2 (Drosophila); polyhomeotic-like protein 2 |
| Gene Symbols | Phc2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.