Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHC3 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077312AF700
Description
PHC3 Polyclonal antibody specifically detects PHC3 in Human samples. It is validated for Western BlotSpecifications
PHC3 | |
Polyclonal | |
Western Blot | |
DKFZp313K1221, early development regulator 3, Early development regulatory protein 3, EDR3FLJ12729, FLJ12967, Homolog of polyhomeotic 3, HPH3, MGC88144, PH3, polyhomeotic homolog 3 (Drosophila), polyhomeotic like 3, polyhomeotic like 3 (Drosophila), polyhomeotic-like protein 3 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human polyhomeotic like 3 using the numbering given in entry AAI31773.1 (GeneID 80012). | |
0.1 mL | |
Chromatin Modifiers | |
80012 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction