Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576951
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
Specifications
PHC3 | |
Polyclonal | |
Unconjugated | |
PHC3 | |
E030046K01Rik; early development regulator 3; early development regulatory protein 3; EDR3; homolog of polyhomeotic 3; hPH3; PH3; Phc3; polyhomeotic 3; polyhomeotic homolog 3; polyhomeotic homolog 3 (Drosophila); polyhomeotic like 3; polyhomeotic-like 3 (Drosophila); polyhomeotic-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
241915, 310258, 80012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CHP6, Q8NDX5 | |
PHC3 | |
Recombinant full length Human PHC3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction