Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111540 Supplier Invitrogen™ Supplier No. PA5111540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSSLLIEQPV KKRPLLDNQV INSVCVQPEL QNNTKHADNS SDTEMEDMIA EETLEEMDSE LLKCEFCGK Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 99%.

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHC3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHC3
Gene Accession No. Q8NDX5
Gene Alias E030046K01Rik; early development regulator 3; early development regulatory protein 3; EDR3; homolog of polyhomeotic 3; hPH3; PH3; Phc3; polyhomeotic 3; polyhomeotic homolog 3; polyhomeotic homolog 3 (Drosophila); polyhomeotic like 3; polyhomeotic-like 3 (Drosophila); polyhomeotic-like protein 3
Gene Symbols PHC3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PHC3. Recombinant protein control fragment (Product #RP-105219).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.