Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538047
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538047 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538047 Supplier Invitrogen™ Supplier No. PA538047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is NIH-3T3 cell lysate. At least six isoforms of PHC3 are known to exist.

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PHC3
Gene Accession No. Q8CHP6, Q8NDX5
Gene Alias E030046K01Rik; early development regulator 3; early development regulatory protein 3; EDR3; homolog of polyhomeotic 3; hPH3; PH3; Phc3; polyhomeotic 3; polyhomeotic homolog 3; polyhomeotic homolog 3 (Drosophila); polyhomeotic like 3; polyhomeotic-like 3 (Drosophila); polyhomeotic-like protein 3
Gene Symbols PHC3
Host Species Rabbit
Immunogen 19 amino acid peptide near the amino terminus of human PHC3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241915, 310258, 80012
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.