Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHC3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549520
Description
Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A ′Lys-119′, rendering chromatin heritably changed in its expressibility.
Specifications
PHC3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NDX5 | |
PHC3 | |
KLH conjugated synthetic peptide between 819-850 amino acids from human PHC3. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PHC3 | |
Polyhomeotic-like protein 3, Early development regulatory protein 3, Homolog of polyhomeotic 3, hPH3, PHC3, EDR3, PH3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80012 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction