Learn More
Invitrogen™ PHD1 Recombinant Rabbit Monoclonal Antibody (2G9M7)

Description
Immunogen sequence: MDSPCQPQPL SQALPQLPGS SSEPLEPEPG RARMGVESYL PCPLLPSYHC PGVPSEASAG SGTPRATATS TTASPLRDGF GGQDGGELRP LQSEGAAALV.
Specifications
Specifications
| Antigen | PHD1 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 2G9M7 |
| Concentration | 0.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EGLN2 |
| Gene Accession No. | Q6AYU4, Q91YE2, Q96KS0 |
| Gene Alias | 0610011A13Rik; C85656; egl nine homolog 2; egl-9 family hypoxia inducible factor 2; egl-9 family hypoxia-inducible factor 2; Egln2; EIT6; EIT-6; Estrogen-induced tag 6; falkor; HIF-1alpha prolyl-4-hydroxylase-1; HIF-P4H 1; Hif-p4h-1; HIFPH1; HIF-PH1; HIF-prolyl hydroxylase 1; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 1; Ier4; immediate early response 4; P4H1; Phd1; PHD-1; Prolyl hydroxylase domain-containing protein 1; SM-20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.