Learn More
Invitrogen™ PHD2 Polyclonal Antibody

Description
Suggested positive control: antigen standard for EGLN1 (transient overexpression lysate).
Specifications
Specifications
| Antigen | PHD2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | Egln1 |
| Gene Accession No. | P59722, Q91YE3, Q9GZT9 |
| Gene Alias | AI503754; C1orf12; C1ORF12 antibody; ECYT3; egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia-inducible factor 1; EGLN 1 EGLN1; EGLN1; HALAH; HIF PH2; HIF prolyl hydroxylase 2; HIF-P4H 2; Hif-p4h-2; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 2; HPH2; HPH-2; Hypoxia-inducible factor prolyl hydroxylase 2; ORF13; P4H2; Phd2; PHD-2; PNAS-118; PNAS-137; Prolyl Hy; Prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6 |
| Gene Symbols | Egln1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.