Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-175 Supplier Invitrogen™ Supplier No. PA116946
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is expected to react with rat based on sequence homology.

SYMPK is a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.This gene encodes a nuclear protein that functions in the regulation of polyadenylation and promotes gene expression. The protein forms a high-molecular weight complex with components of the polyadenylation machinery. It is thought to serve as a scaffold for recruiting regulatory factors to the polyadenylation complex. It also participates in 3'-end maturation of histone mRNAs, which do not undergo polyadenylation. The protein also localizes to the cytoplasmic plaques of tight junctions in some cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHD2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Egln1
Gene Accession No. P59722, Q91YE3, Q9GZT9
Gene Alias AI503754; C1orf12; C1ORF12 antibody; ECYT3; egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia-inducible factor 1; EGLN 1 EGLN1; EGLN1; HALAH; HIF PH2; HIF prolyl hydroxylase 2; HIF-P4H 2; Hif-p4h-2; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 2; HPH2; HPH-2; Hypoxia-inducible factor prolyl hydroxylase 2; ORF13; P4H2; Phd2; PHD-2; PNAS-118; PNAS-137; Prolyl Hy; Prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6
Gene Symbols Egln1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of mouse PHD2/HIF Prolyl Hydroxylase 2 (between residues 300-400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112405, 308913, 54583
Target Species Human, Mouse, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.