Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHD3 Monoclonal Antibody (3C5)
GREENER_CHOICE

Catalog No. PIMA522900
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522900 Supplier Invitrogen™ Supplier No. MA522900
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: EMPLGHIMRL DLEKIALEYI VPCLHEVGFC YLDNFLGEVV GDCVLERVKQ LHCTGALRDG QLAGPRAGVS KRHLRGDQIT WIGGNEEGCE AISFLLSLID.

Cellular oxygen sensor that catalyzes, under normoxic conditions, the post-translational formation of 4-hydroxyproline in hypoxia-inducible factor (HIF) alpha proteins. Hydroxylates a specific proline found in each of the oxygen-dependent degradation (ODD) domains (N-terminal, NODD, and C-terminal, CODD) of HIF1A. Also hydroxylates HIF2A. Has a preference for the CODD site for both HIF1A and HIF2A. Hydroxylation on the NODD site by EGLN3 appears to require prior hydroxylation on the CODD site. Hydroxylated HIFs are then targeted for proteasomal degradation via the von Hippel-Lindau ubiquitination complex. Under hypoxic conditions, the hydroxylation reaction is attenuated allowing HIFs to escape degradation resulting in their translocation to the nucleus, heterodimerization with HIF1B, and increased expression of hypoxy-inducible genes. EGLN3 is the most important isozyme in limiting physiological activation of HIFs (particularly HIF2A) in hypoxia. Also hydroxylates PKM in hypoxia, limiting glycolysis. Under normoxia, hydroxylates and regulates the stability of ADRB2. Regulator of cardiomyocyte and neuronal apoptosis. In cardiomyocytes, inhibits the anti-apoptotic effect of BCL2 by disrupting the BAX-BCL2 complex. In neurons, has a NGF-induced proapoptotic effect, probably through regulating CASP3 activity. Also essential for hypoxic regulation of neutrophilic inflammation. Plays a crucial role in DNA damage response (DDR) by hydroxylating TELO2, promoting its interaction with ATR which is required for activation of the ATR/CHK1/p53 pathway. Target proteins are preferentially recognized via a LXXLAP motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHD3
Applications ELISA
Classification Monoclonal
Clone 3C5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene EGLN3
Gene Accession No. Q9H6Z9
Gene Alias 2610021G09Rik; AI505553; AI648162; Egl nine homolog 3; egl nine homolog 3, mitochondrial; egl nine-like protein 3 isoform; egl-9 family hypoxia inducible factor 3; egl-9 family hypoxia-inducible factor 3; EGLN3; factor-responsive smooth muscle protein; FLJ21620; HIF prolyl hydroxylase 3; HIFP4H3; Hif-p4h-3; HIFPH3; HIF-PH3; HIF-prolyl hydroxylase 3; HPH-1; HPH-3; hypoxia-inducible factor prolyl hydroxylase 3; MGC125998; MGC125999; P4H3; Phd3; PHD-3; prolyl hydroxylase domain-containing protein 3; Sm20; SM-20
Gene Symbols EGLN3
Host Species Mouse
Immunogen EGLN3 (AAH10992.2, 94 a.a. ~ 193 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112399
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.