Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD3 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501557382
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-7382 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-7382 Supplier Bethyl Laboratories, Inc Supplier No. A300327AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

HIF prolyl hydroxylase 3 is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the HPH sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. HPH is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, HPHs convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis. HIF prolyl hydroxylase 1, specifically, catalyzes the posttranslational formation of 4-hydroxyproline in HIF alpha proteins. It hydroxylates HIF-1 alpha at Pro(564) and HIF-2 alpha. It targets HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitylation complex. It may also play a role in cell growth regulation in muscle cells and in apoptosis in neuronal tissues, promoting cell death through a caspase-dependent mechanism.

Specifications

Antigen PHD3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EGLN3
Gene Accession No. Q91UZ4, Q9H6Z9
Gene Alias EGL9 homolog of C. elegans 3, EGLN3, Factor responsive smooth muscle protein, HIF Prolyl Hydroxylase 3, HIFP4H3, HIFPH3, P4H3, Prolyl Hydroxylase Doma
Gene Symbols EGLN3
Host Species Rabbit
Immunogen Between 50 and 100
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112399, 112407
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.