Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHD4/HIF Prolyl Hydroxylase 4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$501.50

Specifications

Antigen PHD4/HIF Prolyl Hydroxylase 4
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100295
SDP
View Documents
Novus Biologicals
NB100295
0.1 mg
Each of 1 for $501.50
Only null left
Add to Cart
 
Description

Description

PHD4/HIF Prolyl Hydroxylase 4 Polyclonal specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications

Specifications

PHD4/HIF Prolyl Hydroxylase 4
Western Blot 1:500-1:2500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy
Polyclonal
Rabbit
Cancer, Hypoxia, Lipid and Metabolism
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase
P4HTM
IgG
Affinity Purified
Human. Does not appear to work in Mouse. Rat reactivity reported in scientific literature (PMID: 17003483).
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
RUO
Human, Rat, Mouse (Negative)
NP_808808.1
54681
The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681).
Primary
Store at 4C. Do not freeze.
50 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.