Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHEMX/ART1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982315
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2315 Supplier Bioss Supplier No. BS12531R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene, which is a member of the tetraspanin superfamily, is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of chromosome 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian and breast cancers. This gene is located among several imprinted genes; however, this gene, as well as the tumor-suppressing subchromosomal transferable fragment 4, escapes imprinting. This gene may play a role in malignancies and diseases that involve this region, and it is also involved in hematopoietic cell function. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.

Specifications

Antigen PHEMX/ART1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TSPAN32
Gene Accession No. Q96QS1
Gene Alias AML1-regulated transmembrane protein 1; Art1; Art-1; AW208513; BB235973; D7Wsu37e; FLJ17158; FLJ97586; MGC22455; pan hematopoietic expression; pan-hematopoietic expression protein; Phemx; PHMX; Protein Phemx; tetraspanin; tetraspanin 32; Tetraspanin-32; TSPAN32; Tspan-32; Tssc6; tumor-suppressing STF cDNA 6; tumor-suppressing subchromosomal transferable fragment 6; tumor-suppressing subchromosomal transferable fragment cDNA 6; tumor-suppressing subtransferable candidate 6
Gene Symbols TSPAN32
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TSPAN32.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10077
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.