Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenamil Methanesulfonate, Thermo Scientific™

Catalog No. AC329300050
Click to view available options
Quantity:
5 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1161-94-0
Molecular Formula C13H16ClN7O4S
Molecular Weight (g/mol) 401.826
MDL Number MFCD00274070
InChI Key MHPIZTURFVSLTJ-UHFFFAOYSA-N
Synonym phenamil methanesulfonate, phenamil methanesulfonate salt, 3,5-diamino-6-chloro-n-imino phenylamino methyl pyrazinecarboxamide methanesulfonate salt, lopac-p-203, phenamil methanesulfonate salt, solid, phenamil methanesulfonate free base, 3,5-diamino-6-chloro-n-n-phenylcarbamimidoyl pyrazine-2-carboxamide methanesulfonate, 3,5-diamino-6-chloro-n-imino phenylamino methyl-pyrazinecarboxamide methansulfonate
PubChem CID 9604979
IUPAC Name 3,5-diamino-N-[amino(anilino)methylidene]-6-chloropyrazine-2-carboxamide;methanesulfonic acid
SMILES CS(=O)(=O)O.C1=CC=C(C=C1)NC(=NC(=O)C2=C(N=C(C(=N2)Cl)N)N)N

Specifications

Melting Point 255°C to 263°C
Color White to Yellow
Infrared Spectrum Authentic
Quantity 5 mg
Formula Weight 401.8
Physical Form Solid
Chemical Name or Material Phenamil methanesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.