Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazine, 99.2%, MP Biomedicals™

Catalog No. ICN10260501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10260501 1 g
ICN10260510 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10260501 Supplier MP Biomedicals, Inc Supplier No. 0210260501
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-82-0
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
InChI Key PCNDJXKNXGMECE-UHFFFAOYSA-N
Synonym dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
PubChem CID 4757
ChEBI CHEBI:36674
IUPAC Name phenazine
SMILES C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

Specifications

Color Yellow
Assay Percent Range 99.2%
Quantity 1 g
Formula Weight 180.2
Physical Form Crystalline Powder
Chemical Name or Material Phenazine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.