Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl isocyanate, 99%

Catalog No. p-3213038
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

CAS: 1943-82-4 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1943-82-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00037044
InChI Key HACRKYQRZABURO-UHFFFAOYSA-N
Synonym phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate
PubChem CID 160602
ChEBI CHEBI:60067
IUPAC Name 2-isocyanatoethylbenzene
SMILES C1=CC=C(C=C1)CCN=C=O

Specifications

Color Colorless to Yellow
Density 1.0630g/mL
Boiling Point 210°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5210 to 1.523
Linear Formula C6H5(CH2)2NCO
Quantity 5 mL
Specific Gravity 1.063
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene
Formula Weight 147.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.