Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, 99.5%, extra pure, loose crystals, unstabilized

Catalog No. ac221750015
Click to view available options
Quantity:
1.5 kg

CAS: 108-95-2 | C6H6O | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39.0°C to 42.0°C
Density 1.0700g/mL
Boiling Point 182.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Linear Formula C6H5OH
Quantity 1.5 kg
Beilstein 06,11
Residue after Evaporation 0.05% max.
Fieser 01,828; 13,166; 14,88
Merck Index 15,7354
Specific Gravity 1.07
Solubility Information Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
Viscosity 3.437 mPa.s (50°C)
Formula Weight 94.11
Percent Purity 99.5%
Grade Extra Pure
Chemical Name or Material Phenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 203-632-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.