Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol (Crystals), Fisher Chemical™

Catalog No. A92112 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
1.5 kg
Packaging:
Amber Glass/Poison Pack

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39°C
Color Colorless
Density 1.070g/cm³
pH 4 to 5
Boiling Point 182°C
Flash Point 79°C
Assay Percent Range ≥99.0 %
Packaging Amber Glass/Poison Pack
Linear Formula C6H5OH
Quantity 1.5 kg
UN Number 1671
Residue after Evaporation 0.05% max.
Viscosity 3.437 mPa/s (50°C)
Vapor Pressure 0.4mbar at 20°C
Formula Weight 94.11
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material Phenol
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible material. Causes burns by all exposure routes. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:2
Instability:1
DOTInformation DOT Class 6.1, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.