Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol, Fused Crystal, USP, 99-100.5%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
500 g
1 kg
1.5 kg
6 x 500 g
25 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Phenol, Fused Crystal, USP is extracted from petroleum and mainly used to produce precursors of plastics and is also used as a building block to synthetically produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesic among them. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Percent Purity | 99 to 100.5% |
Grade | USP |
Packaging | Amber Glass Bottle |
Color | Colorless |
Quantity | 1.5 kg |
Physical Form | Fused Crystal |
Chemical Name or Material | Phenol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction