Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol, Loose Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18605080
Click to view available options
Quantity:
125 g
500 g
1 kg
6 x 125 g
6 x 500 g
4 x 1 kg
45 kg
Packaging:
Amber Glass Bottle

P1060, 108-95-2, C6H6O

Spectrum™ Chemical Phenol, Loose Crystal, Reagent, ACS is a white crystalline, aromatic organic compound that is volatile. It's extracted from petroleum and mainly used as a synthetic building block to produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 1
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Color Colorless to White
Quantity 6 x 500 g
Physical Form Loose Crystals
Chemical Name or Material Phenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.