Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenol Red, ACS Reagent (Phenolsulfonphthalein), Reagents

Catalog No. 01336731
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-731 5 g
01-336-732 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-731 Supplier Reagents Holdings Llc Supplier No. C61225005C2
Add to Cart
Add to Cart

Phenol Red, ACS Reagent (Phenolsulfonphthalein), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Boiling Point 563°C
Grade ACS
Packaging Glass Bottle
Specific Gravity 1
Color Red
Quantity 5 g
Physical Form Solid
Chemical Name or Material Phenol Red, ACS Reagent (Phenolsulfonphthalein)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.