Learn More
Thermo Scientific Chemicals Phenol Red, ACS
Description
Phenol red is used as non absorbed marker in the gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >300°C |
| Beilstein | 326470 |
| Merck Index | 14,7247 |
| Solubility Information | Practically insoluble in chloroform,ether; Soluble in aqueous alkali hydroxides or carbonates with red color (removed by boiling with zinc dust); Solubility in water: approximately 1g/1300ml; Solubility in alcohol: approximately 1g/350mlSoluble in methanol. |
| IUPAC Name | 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| Formula Weight | 354.38 |
| Grade | ACS |
| Odor | Odorless |
| Quantity | 5 g |
| Physical Form | Crystalline |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 205-609-7 |
| RTECSNumber | SJ7490000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.