Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Phenol Red Indicator
SCI_ED

Catalog No. S93322 Shop All Aldon Products
Click to view available options
Quantity:
10 g

Test water pH within 6.8-8.2 range with this phenol red indicator.

Turns from yellow to red to bright pink as pH increases.

Specifications

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Linear Formula C19H13NaO5S
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
Molecular Weight (g/mol) 376.358
PubChem CID 23686673
Formula Weight 376.36
Percent Purity 99%
Grade Reagent
Packaging ACS Grade - Sodium Salt
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
pH Color Changes pH 6.8-8.2 (yellow-red)
Color Red
Quantity 10 g
Chemical Name or Material Phenol Red Indicator
Show More Show Less
ShelfLife 36 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.