Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phenol Red sodium salt, ACS

Catalog No. p-7049609
Click to view available options
Quantity:
5 g
25 g

pH indicator | CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

Phenol red sodium salt is a pH indicator commonly used in cell culture for monitoring pH changes. It is used in the pH ranges between 6.8 and 8.2. It is involved in the studies of gastrointestinal and other systems. It can be used to check hydrogen peroxide in supernatants from cultured macrophages in multiwell plates. Further, it is used as a diagnostic aid for determination of renal function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285°C (decomposition)
Beilstein 6260026
Merck Index 14,7247
Solubility Information Soluble in water,alcohol,aqueous alkali hydroxides and carbonates.
Formula Weight 376.37
Grade ACS
Quantity 5 g
Physical Form Crystalline
Chemical Name or Material Phenol Red sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-057-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.