Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenol Red TS, 0.1% (w/v) Alcoholic, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | Phenol Red |
Name Note | Test Solution |
CAS | 64-17-5, 143-74-8, 67-56-1 |
CAS Min % | 98.0, 0.12 |
CAS Max % | 100.0, 0.13 |
Concentration | 0.1% (w/v) |
Grade | Indicator |
Packaging | Amber Glass Bottle |
Solvent or Matrix | Alcoholic Solution |
Color | Orange/Red |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction