Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, 0.2% (w/v) in denatured Ethanol, Ricca Chemical
SDP

Catalog No. R5597000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R5597000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5597000500 Supplier Ricca Chemical Company Supplier No. R5597000500A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 77-09-8
Molecular Formula C2H6O
Molecular Weight (g/mol) Mixture
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 95, 4.75, 0.25
Boiling Point Approximately 77°C
Packaging Natural Poly Bottle
Quantity 500 mL
Percent Purity 0.2% w/v
Grade Laboratory
Physical Form Liquid
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.