Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, 1% w/v in Methanol, Reagents

Catalog No. 01334396
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-396 4 L
01-334-395 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-396 Supplier Reagents Holdings Llc Supplier No. CP1078004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenolphthalein, 1% w/v in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 77-09-8 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 65°C
Packaging Poly Bottle
Specific Gravity 0.783
Color Colorless
Quantity 4 L
Physical Form Liquid
Percent Purity 1%
Grade Reagent
Chemical Name or Material Phenolphthalein, 1% w/v in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.