Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phenolphthalein, ACS

Catalog No. p-7050575
Click to view available options
Quantity:
100 g
500 g

Indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extend of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258°C to 262°C
Beilstein 284423
Merck Index 14,7243
Solubility Information Soluble in alcohol; Slightly soluble in ether; Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
Formula Weight 318.33
Grade ACS
Density 1.299 g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Phenolphthalein, For analysis ACS
Hazard Category H341-H351-H361f-H373
Hazard Statement GHS H Statement
H350-H341-H361
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 201-004-7
RTECSNumber SM8380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.