Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phenolphthalein Indicator, 1% (w/v) in 95% (v/v) Alcohol, Neutralized, Ricca Chemical

Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | Phenolphthalein Indicator |
CAS | 64-17-5, 7732-18-5, 67-56-1, 77-09-8 |
Percent Purity | 1% W/V |
Grade | Indicator |
Concentration or Composition (by Analyte or Components) | 1% (w/v) |
Packaging | Natural Poly Bottle |
Solvent or Matrix | 95% (v/v) Alcohol |
Color | Colorless to Pink |
Quantity | 4 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction