Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Indicator, 0.08% (w/v) in Methanol, Ricca Chemical
SDP

Catalog No. p-2916777 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle

Catalog No. 5595-4

Supplier: Ricca Chemical Company 55954

Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Phenolphthalein Indicator
CAS 67-56-1, 7732-18-5, 77-09-8
Percent Purity 0.08% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.08% (w/v) in Methanol
Packaging Natural Poly Bottle
Color Colorless to Pink
Quantity 120 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.