Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Phenolphthalein in IPA Solution
Shop All Fisherbrand Products

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
0.5%
1%
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein in IPA Solution |
| CAS | 77-09-8, 7732-18-5 |
| Packaging | 0.5% |
| Density | 0.85 to 0.95 |
| Color | Colorless to Pink |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction