Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, Powder, Reagent, ACS, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

P1075, 77-09-8, C20H14O4

Spectrum™ Phenolphthalein, Powder, Reagent, ACS is a chemical compound that varies in color according to the acidity level of the solution it is in. It has traditionally been used as a laxative but has more recently been used as part of a test to identify blood in substances, known as the Kastle-Mayer test. It is also used in certain toys to create disappearing ink effect. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Packaging Poly Bottle
Quantity 25 g
CAS Max % 1
Grade ACS