Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein, Reagent ACS Meets USP and ACS Specifications, Reagents

Catalog No. 01337133
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-133 500 g
01-337-137 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-133 Supplier Reagents Holdings Llc Supplier No. C2270000500A5
Add to Cart
Add to Cart

Phenolphthalein, Reagent ACS Meets USP and ACS Specifications, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 258°C to 262°C
Boiling Point 558°C
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.299
pH Color Changes Colorless (pH 8.0) to Pink (pH 10.0)
Color White to Yellow
Quantity 500 g
Physical Form Solid
Chemical Name or Material Phenolphthalein, Reagent ACS Meets USP and ACS Specifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.