Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein Stock Solution, Fisher Chemical™
SDP

Catalog No. p-213683 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 mL
50 mL
Packaging:
Natural Poly Bottle

Specifications

Chemical Name or Material Phenolphthalein Stock Solution
CAS 7732-18-5, 64-17-5, 67-56-1, 67-63-0, 77-09-8
CAS Min % 52.0, 43.0, 2.0, 2.0, 0.01
CAS Max % 55.0, 44.0, 3.0, 3.0, 0.01
Grade Indicator
Packaging Natural Poly Bottle
Color Colorless
pH 8
Quantity 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.