Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolphthalein TS, Reagents

Catalog No. 01334113
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
4 L

Catalog No. 01-334-113

Supplier: Reagents Holdings Llc CP2607001A

Only null left
Add to Cart
Add to Cart

Phenolphthalein TS, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5, 7732-18-5, 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 77°C
Grade USP
Packaging Poly Bottle
Specific Gravity 0.7861
Color Colorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Phenolphthalein TS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.