Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenolsulfonic Acid, 65%, Spectrum™ Chemical
SDP

Catalog No. 18605099
Click to view available options
Quantity:
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum

PH136, 1333-39-7, HOC6H4SO3H

Spectrum™ Chemical Phenolsulfonic Acid

Chemical Identifiers

CAS 1333-39-7
Molecular Formula C6H6O4S
Molecular Weight (g/mol) 174.17
InChI Key IULJSGIJJZZUMF-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzene-1-sulfonic acid
SMILES OC1=CC=CC=C1S(O)(=O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 65%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.