Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol-d4, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D
Click to view available options
Quantity:
7.5 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Specifications
| Physical Form | Liquid |
| Flash Point | 12°C |
| Color | Colorless |
| Refractive Index | 1.3262 |
| Quantity | 7.5 mL |
| Deuterium (D) | 99.95% min. |
| Density | 0.8880g/mL |
| Water | 0.02% max. (Coulometric) |
| Melting Point | -99.0°C |
| Boiling Point | 65.0°C |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes damage to organs. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Immediately call a POISON CENTER or doctor/physician. IF exposed: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction