Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenothiazine, 98+%

Catalog No. aaa1251730
Click to view available options
Quantity:
250 g
500 g
5000 g

Catalog No. AAA1251730

Supplier: Thermo Scientific Chemicals A1251730

Only null left
Add to Cart

CAS: 92-84-2 | C12H9NS | 199.271 g/mol

It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.

Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.

Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-84-2
Molecular Formula C12H9NS
Molecular Weight (g/mol) 199.271
MDL Number MFCD00005015
InChI Key WJFKNYWRSNBZNX-UHFFFAOYSA-N
Synonym phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazine
PubChem CID 7108
ChEBI CHEBI:37931
IUPAC Name 10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=CC=CC=C3S2

Specifications

Melting Point 182°C to 186°C
Boiling Point 235°C (21 mmHg)
Quantity 250 g
Beilstein 143237
Sensitivity Light sensitive
Merck Index 14,7252
Solubility Information Soluble in water (0.13mg/ml at 23°C),ethanol,acetone,and ether. Insoluble in chloroform.
Formula Weight 199.28
Percent Purity ≥98%
Chemical Name or Material Phenothiazine
Hazard Category H302-H317-H373
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 202-196-5
RTECSNumber SN5075000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.